Iron(III) oxide, 98% (metals basis), Thermo Scientific Chemicals
Iron(III) oxide, 98% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(III) oxide, 98% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012375.A1
also known as 012375-A1
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.30 
Save 9.65 (15%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.30 
Save 9.65 (15%)
Add to cart
Iron(III) oxide, 98% (metals basis), Thermo Scientific Chemicals
Catalog number012375.A1
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.30 
Save 9.65 (15%)
-
Add to cart
Chemical Identifiers
CAS1309-37-1
IUPAC Nametrioxodiiron
Molecular FormulaFe2O3
InChI KeyJEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES[Fe][Fe](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities2% max.
It has many applications, mainly used in iron industry, pigment, as photocatalyst, medicine and polishing jewels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only