Gadolinium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Gadolinium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gadolinium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 012396.FI
also known as 012396-FI
Price (USD)/ Each
347.65
Online exclusive
386.00 
Save 38.35 (10%)
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
347.65
Online exclusive
386.00 
Save 38.35 (10%)
Add to cart
Gadolinium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012396.FI
Price (USD)/ Each
347.65
Online exclusive
386.00 
Save 38.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-54-2
IUPAC Namegadolinium
Molecular FormulaGd
InChI KeyUIWYJDYFSGRHKR-UHFFFAOYSA-N
SMILES[Gd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Oxide of Gadolinium foil is used in the control rods of some nuclear reactors. It is used as nuclear control rod material. Neutron shielding, phosphor activator, catalyst, scavenger for oxygen in titanium production. A metal element that is used in magnetic resonance imaging (MRI) and other imaging methods. It has been used in making gadolinium yttrium garnets, which have microwave applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only