Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals
Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 012397.FF
also known as 012397-FF
Price (USD)/ Each
167.00
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
167.00
Add to cart
Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012397.FF
Price (USD)/ Each
167.00
-
Add to cart
Chemical Identifiers
CAS7440-54-2
IUPAC Namegadolinium
Molecular FormulaGd
InChI KeyUIWYJDYFSGRHKR-UHFFFAOYSA-N
SMILES[Gd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only