Lutetium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Lutetium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lutetium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 012410.03
also known as 012410-03
Price (USD)/ Each
228.65
Online exclusive
254.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
228.65
Online exclusive
254.00 
Save 25.35 (10%)
Add to cart
Lutetium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012410.03
Price (USD)/ Each
228.65
Online exclusive
254.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7439-94-3
IUPAC Namelutetium
Molecular FormulaLu
InChI KeyOHSVLFRHMCKCQY-UHFFFAOYSA-N
SMILES[Lu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only