Thermo Scientific Chemicals

1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals

Catalog number: 012411.B4
16 kg, Each
Thermo Scientific Chemicals

1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals

Catalog number: 012411.B4
16 kg, Each
Quantity
Catalog number: 012411.B4
also known as 012411-B4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-63-6
IUPAC Name
1,2,4-trimethylbenzene
Molecular Formula
C9H12
InChI Key
GWHJZXXIDMPWGX-UHFFFAOYSA-N
SMILES
CC1=CC=C(C)C(C)=C1
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless

Description

1,2,4-Trimethylbenzene is applied as a liquid scintillator. It is also used as a sterilizing agent and in the manufacture of dyes, perfumes, and resins. It is used as a safe solvent for the production of H2O2 (hydrogen peroxide). Pseudocumene is used mainly as raw material for the production of trimellitic anhydride, pyromellitic dianhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text