Scandium foil, 0.025mm (0.001in) thick, 99% (REO), Thermo Scientific Chemicals
Scandium foil, 0.025mm (0.001in) thick, 99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Scandium foil, 0.025mm (0.001in) thick, 99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 012426.FI
Price (USD)/ Each
1,853.00
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
1,853.00
Add to cart
Scandium foil, 0.025mm (0.001in) thick, 99% (REO), Thermo Scientific Chemicals
Catalog number012426.FI
Price (USD)/ Each
1,853.00
-
Add to cart
Chemical Identifiers
CAS7440-20-2
IUPAC Namescandium
Molecular FormulaSc
InChI KeySIXSYDAISGFNSX-UHFFFAOYSA-N
SMILES[Sc]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
It finds applications in the aerospace industry. Another use of Scandium is in aluminum-scandium alloys for sports equipment, such as bicycle frames, lacrosse sticks, and baseball bats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only