Terbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Terbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Terbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Form: Approx. 0.52g/25 x 25mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 012430.FF
also known as 012430-FF
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
Add to cart
Terbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012430.FF
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-27-9
IUPAC Nameterbium
Molecular FormulaTb
InChI KeyGZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES[Tb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only