Thermo Scientific Chemicals

Copper(II) chloride dihydrate, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 012458.22
100 g, Each
Thermo Scientific Chemicals

Copper(II) chloride dihydrate, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 012458.22
100 g, Each
Quantity
Catalog number: 012458.22
also known as 012458-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10125-13-0
IUPAC Name
copper(2+) dihydrate dichloride
Molecular Formula
Cl2CuH4O2
InChI Key
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Cu++]
Assay (unspecified)
≥99.0% CuCl₂•2H₂O
Impurity content
Ca: ≤0.005%
Impurity content
Fe: ≤0.005%
Impurity content
K: ≤0.01%
Impurity content
Na: ≤0.02%

Description

Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text