Gallium Indium eutectic, 99.99% (metals basis), Thermo Scientific Chemicals
Gallium Indium eutectic, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gallium Indium eutectic, 99.99% (metals basis), Thermo Scientific Chemicals

Gallium Indium eutectic, Ga:In; 75.5:24.5 wt%, 50 g, 99.99% (metals basis), 6.2, Hazard Class: 8; Packaging Group: III, 231-163-8, H290-H314-H335, P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c, 15.7 deg.C, MFCD00144387, Liquid
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
50 g
Catalog number 012478.09
also known as 012478-09
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
Add to cart
Gallium Indium eutectic, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number012478.09
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGallium Indium eutectic
Melting Point15.7°C
Health Hazard 1H290-H314-H335
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
Recommended StorageAmbient temperatures
View more
It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices. Gallium-Indium eutectic top contact, and a halogen light source was used to test P3HT-PCBM organic solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only