Thermo Scientific Chemicals

Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 012482.22
100 g, Each
Thermo Scientific Chemicals

Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 012482.22
100 g, Each
Quantity
Catalog number: 012482.22
also known as 012482-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12070-12-1
IUPAC Name
methanidylidynetungstenylium
Molecular Formula
CW
InChI Key
UONOETXJSWQNOL-UHFFFAOYSA-N
SMILES
[C-]#[W+]
Assay (unspecified)
C 5.9% min
Total Metal Impurities
0.5% max.
Assay (unspecified)
W 91% min

Description

Tungsten carbide is extensively used as a cutting tool for machining and in armor-piercing ammunition. It is also used in farriery, the shoeing of horses as well as to improve traction on slippery surfaces like roads or ice. It finds application in nuclear reactors as an effective neutron reflector. It is also used for making surgical instruments meant for open surgery such as scissors, forceps, hemostats, blade-handles and in laparoscopic surgery instruments such as graspers, cutter, needle holder and cautery. Further, it also used to make the rotating ball in the tips of ballpoint pens, bridal jewelry, and gauge blocks, which is useful for producing precision lengths in dimensional metrology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text