Thermo Scientific™

Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals

Catalog number: 012483.22
100 g, Each
Thermo Scientific™

Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals

Catalog number: 012483.22
100 g, Each
Quantity
Catalog number: 012483.22
also known as 012483-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1309-60-0
IUPAC Name
plumbanedione
Molecular Formula
O2Pb
InChI Key
YADSGOSSYOOKMP-UHFFFAOYSA-N
SMILES
O=[Pb]=O
Impurity content
Carbon compounds (as C) 0.04%
Impurity content
Na 0.1%
Assay (unspecified)
97.0% PbO₂ min
Impurity content
Cl 0.002%
Impurity content
Mn 5ppm

Description

It finds applications as an electrode in lead acid batteries, as anode in electroplating copper and zinc in sulfate solution, production of glyoxylic acid, manufacturing of pigments and dyes, high voltage lightening arrestors, and as an analytical lab reagent. Lead dioxide is a potent oxidizing agent which is employed in the manufacture of matches, pyrotechnics and other chemicals. It can be used in the curing of sulfide-based polymers. It oxidizes Mn(II) into Mn(VII), and Cr(III) into Cr(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Acid-insoluble matter 0.2%, Carbon compounds (as C) 0.04%, Cl 0.002%, NO3 0.02%, SO4 0.05%, Mn 5ppm, Ca 0.02%, Cu 0.05%, K 0.05%, Na 0.1%
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text