Thermo Scientific Chemicals

Iron(II) chloride hydrate, Reagent Grade, 99%, Thermo Scientific Chemicals

Catalog number: 012484.A7
5 kg, Each
Thermo Scientific Chemicals

Iron(II) chloride hydrate, Reagent Grade, 99%, Thermo Scientific Chemicals

Catalog number: 012484.A7
5 kg, Each
Quantity
Catalog number: 012484.A7
also known as 012484-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23838-02-0
IUPAC Name
λ²-iron(2+) dichloride
Molecular Formula
Cl2Fe . x H2O
InChI Key
NMCUIPGRVMDVDB-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Fe++]
Assay (unspecified)
98.6% min

Description

It is used as a reducing agent in metallurgy, in pharmaceutical preparations, as a mordant in dyeing and in sewage treatment. Ferrous chloride has a kind of niche application. During the laboratory synthesis of iron complexes, ferrous chloride acts as a reducing flocculating agent in wastewater treatment, particularly for wastes containing chromate. It is the precursor to hydrated iron(III) oxides that are magnetic pigments. Ferrous chloride is employed as a reducing agent in many organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text