Thermo Scientific Chemicals

Strontium chloride hexahydrate, tech., Thermo Scientific Chemicals

Catalog number: 012494.A3
2 kg, Each
Thermo Scientific Chemicals

Strontium chloride hexahydrate, tech., Thermo Scientific Chemicals

Catalog number: 012494.A3
2 kg, Each
Quantity
Catalog number: 012494.A3
also known as 012494-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-70-4
IUPAC Name
strontium(2+) hexahydrate dichloride
Molecular Formula
Cl2H12O6Sr
InChI Key
AMGRXJSJSONEEG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Form
Powder or crystals or chunks
Appearance (Color)
White
Assay from Supplier's CofA
Strontium: 30.0-35.0%
Total Metal Impurities
≤5.0%

Description

Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the field of medicine, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text