Strontium oxide, tech., Thermo Scientific Chemicals
Strontium oxide, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium oxide, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 012495.A1
also known as 012495-A1
Price (USD)/ Each
379.00
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
379.00
Add to cart
Strontium oxide, tech., Thermo Scientific Chemicals
Catalog number012495.A1
Price (USD)/ Each
379.00
-
Add to cart
Chemical Identifiers
CAS1314-11-0
IUPAC Namestrontium(2+) oxidandiide
Molecular FormulaOSr
InChI KeyIATRAKWUXMZMIY-UHFFFAOYSA-N
SMILES[O--].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Impurity contentStrontium carbonate: ≤3.0%
Appearance (Color)White to gray
Assay from Supplier's CofA≥96.0%
In the manufacture of strontium salts, pyrotechnics, pigments, in greases and soaps and as a desiccant.Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industries. It is an active component of paint dryers. It is also used to prepare pure strontium upon heating with aluminum under vacuum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant
RUO – Research Use Only