Iron(III) chloride hexahydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals
Iron(III) chloride hexahydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(III) chloride hexahydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals

Used to prepare organically templated iron phosphates.Also, used for sewerage treatment of industrial wastes, chloride hydrometallurgy. Useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1 kg
5 kg
Catalog number 012497.30
also known as 012497-30
Price (USD)/ Each
46.65
Online exclusive
52.00 
Save 5.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
52.00 
Save 5.35 (10%)
Add to cart
Iron(III) chloride hexahydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals
Catalog number012497.30
Price (USD)/ Each
46.65
Online exclusive
52.00 
Save 5.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10025-77-1
IUPAC Nameiron(3+) hexahydrate trichloride
Molecular FormulaCl3FeH12O6
InChI KeyNQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLumps, deliquescent solid
Impurity contentFerrous iron: ≤0.002%
Impurity contentMagnesium (Mg): ≤0.005%
Impurity contentNitrate(NO₃): ≤0.01%
Impurity contentPhosphorus compounds (as PO₄): ≤0.01%
View more
Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications:
  • Used to prepare organically templated Iron Phosphates
  • Used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin) process for production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards
  • Plays important role in photoengraving and photography as an etching agent
  • Usedl as mordant in dyeing and printing textiles.
  • Also usedl in detection of phenols and phenol derivatives; gamma-hydroxybutyric acids, in Trinder spot test for detecting salicylic acids
RUO – Research Use Only