Thermo Scientific Chemicals

Chromium(III) acetate hydroxide, Cr 24%, Thermo Scientific Chemicals

Catalog number: 012518.36
500 g, Each
Thermo Scientific Chemicals

Chromium(III) acetate hydroxide, Cr 24%, Thermo Scientific Chemicals

Catalog number: 012518.36
500 g, Each
Quantity
Catalog number: 012518.36
also known as 012518-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39430-51-8
IUPAC Name
chromium(3+) triacetate
Molecular Formula
C6H9CrO6
InChI Key
WYYQVWLEPYFFLP-UHFFFAOYSA-K
SMILES
[Cr+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Assay (unspecified)
Cr 24% min

Description

Occasionally to dehalogenate organic compounds. used as an oxygen scrubber, polymer industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text