Thermo Scientific Chemicals

Antimony(III) sulfide, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 012521.30
250 g, Each
Thermo Scientific Chemicals

Antimony(III) sulfide, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 012521.30
250 g, Each
Quantity
Catalog number: 012521.30
also known as 012521-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1345-04-6
IUPAC Name
diantimony(3+) trisulfanediide
Molecular Formula
S3Sb2
InChI Key
NVWBARWTDVQPJD-UHFFFAOYSA-N
SMILES
[S--].[S--].[S--].[Sb+3].[Sb+3]
Total Metal Impurities
2% max.

Description

It is manufactured for use in safety matches, military ammunition, explosives and fireworks. It also is used in the production of ruby-colored glass and in plastics as a flame retardant. percussion caps, camouflage paints, ruby glass

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text