Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Thermo Scientific Chemicals
Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Thermo Scientific Chemicals

For Use As a Catalyst, Nitrating Agent For Aromatic organosilicon Compounds, Nickel-Plating Baths
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
50 g
500 g
2 kg
Catalog number 012523.A9
also known as 012523-A9
Price (USD)/ Each
980.65
Online exclusive
1089.00 
Save 108.35 (10%)
-
Add to cart
Quantity:
10 kg
Request bulk or custom format
Price (USD)/ Each
980.65
Online exclusive
1089.00 
Save 108.35 (10%)
Add to cart
Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Thermo Scientific Chemicals
Catalog number012523.A9
Price (USD)/ Each
980.65
Online exclusive
1089.00 
Save 108.35 (10%)
-
Add to cart
Chemical Identifiers
CAS19004-19-4
IUPAC Namedicopper(2+) pentahydrate tetranitrate
Molecular FormulaCu2H10N4O17
InChI KeyIZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCl: ≤0.002%
Impurity contentFe: ≤0.005%
Impurity contentK: ≤0.005%
Impurity contentNa: ≤0.01%
Impurity contentNi: ≤0.01%
View more
Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black and producing a burnished effect on ironCopper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only