Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals
Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number 012524.36
also known as 012524-36
Price (USD)/ Each
240.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
240.00
Add to cart
Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number012524.36
Price (USD)/ Each
240.00
-
Add to cart
Chemical Identifiers
CAS21041-93-0
IUPAC Nameλ²-cobalt(2+) dihydroxide
Molecular FormulaCoH2O2
InChI KeyASKVAEGIVYSGNY-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Cobalt(II) hydroxide is used as a drying agent for paints and varnishes. It acts as an aid to dry the lithographic inks and catalyst in the preparation of battery electrode impregnating solutions. It is also involved in the synthesis of other cobalt compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only