Thermo Scientific Chemicals

Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals

Catalog number: 012527.30
250 g, Each
Thermo Scientific Chemicals

Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals

Catalog number: 012527.30
250 g, Each
Quantity
Catalog number: 012527.30
also known as 012527-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14363-15-6
IUPAC Name
zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate
Molecular Formula
C10H16O5Zn
InChI Key
BNHYOPPPENBGNL-SUKNRPLKSA-L
SMILES
O.[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=[O-]
Appearance (Color)
White to pale cream
Insoluble material
≤0.30%
Water Content (Karl Fischer Titration)
≤11.0%

Description

Zinc Metal acetylacetonates are used as catalysts for polymerization of olefins and transesterification. It is used as a a stabilizer PVC.They are used as solvents, lubricant additives, paint drier, and pesticides. They are used in glass coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text