Thermo Scientific Chemicals

Iron(III) 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog number: 012534.22
100 g, Each
Thermo Scientific Chemicals

Iron(III) 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog number: 012534.22
100 g, Each
Quantity
Catalog number: 012534.22
also known as 012534-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14024-18-1
IUPAC Name
iron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C15H21FeO6
InChI Key
AQBLLJNPHDIAPN-LNTINUHCSA-K
SMILES
[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Assay (unspecified)
Volatile content: 0.30%, max
Insoluble material
0.30 wt%, max
Melting Point
170 to 190 deg C
Form
powder
Appearance (Color)
Red

Description

Iron(III) 2,4-pentanedionate is used as a precatalyst and reagent in organic chemistry. It acts as a precursor for highly crystalline (Zn, Fe)Fe2O4 films and magnetic property measurements of these films. It catalyzes the dimerization of isoprene to a mixture of 1,5-dimethyl-1,5-cyclooctadiene and 2,5-dimethyl-1,5-cyclooctadiene, ring-opening polymerization of 1,3-benzoxazine and catalyze the reaction of N-sulfonyl oxaziridines with olefins to form 1,3-oxazolidine products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text