Thermo Scientific Chemicals

Cobalt(III) 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog number: 012536.22
100 g, Each
Thermo Scientific Chemicals

Cobalt(III) 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog number: 012536.22
100 g, Each
Quantity
Catalog number: 012536.22
also known as 012536-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21679-46-9
IUPAC Name
2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5⁶.5⁶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide
Molecular Formula
C15H21CoO6
InChI Key
VACIXPOMJJIDBM-UHFFFAOYSA-N
SMILES
CC1=[O][Co+3]234([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2.CC([CH-]C(C)=[O]3)=[O]4
Form
Crystals
Assay from Suppliers CofA
Cobalt (Co) content: 16.3-17.0%
Insoluble material
≤1.0% (from solution in Dichloromethane)
Volatiles
≤1.0%
Appearance (Color)
Dark green to black

Description

Cobalt(III) 2,4-pentanedionate is used as a polymerization catalyst, vulcanizing agent, pigment in resins and glass. It is also used for the preparation of monoporphyrinates. Further, it is used in kinetic studies of iron-sulfur proteins. In addition to this, it reacts with trimethylaluminum to prepare Me(3Al/Co(acac)3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text