Cobalt(II) 2,4-pentanedionate, Thermo Scientific Chemicals
Cobalt(II) 2,4-pentanedionate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) 2,4-pentanedionate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 012537.18
also known as 012537-18
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
Add to cart
Cobalt(II) 2,4-pentanedionate, Thermo Scientific Chemicals
Catalog number012537.18
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
-
Add to cart
Chemical Identifiers
CAS14024-48-7
IUPAC Nameλ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14CoO4
InChI KeyUTYYEGLZLFAFDI-FDGPNNRMSA-L
SMILES[Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)21.0% Cobalt, min
Comment0.3% max, percent volatile
Insoluble material1% max
Cobalt(II) 2,4-pentanedionate is used as an important catalyst for organic reactions. It is also used as paint dryer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only