Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals
Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 012542.22
also known as 012542-22
Price (USD)/ Each
37.10
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
37.10
Add to cart
Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals
Catalog number012542.22
Price (USD)/ Each
37.10
-
Add to cart
Chemical Identifiers
CAS13963-57-0
IUPAC Namealuminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21AlO6
InChI KeyKILURZWTCGSYRE-LNTINUHCSA-K
SMILES[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay from Suppliers CofAInsoluble material: ≤0.30%
Assay from Suppliers CofAVolatile material: ≤0.30%
Melting Point (clear melt)188-197°C
Alumunium acetylacetonate may be used to prepare transparent superhydrophobic boehmite and silica films by sublimation, to deposit alumunium oxide films by chemical vapor deposition, to deposit alumunium oxide films by chemical vapor deposition, as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only