Thermo Scientific Chemicals

Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog number: 012542.36
500 g, Each
Thermo Scientific Chemicals

Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog number: 012542.36
500 g, Each
Quantity
Catalog number: 012542.36
also known as 012542-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13963-57-0
IUPAC Name
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C15H21AlO6
InChI Key
KILURZWTCGSYRE-LNTINUHCSA-K
SMILES
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
Insoluble material: ≤0.30%
Assay from Suppliers CofA
Volatile material: ≤0.30%
Melting Point (clear melt)
188-197°C

Description

Alumunium acetylacetonate may be used to prepare transparent superhydrophobic boehmite and silica films by sublimation, to deposit alumunium oxide films by chemical vapor deposition, to deposit alumunium oxide films by chemical vapor deposition, as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text