Indium(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Indium(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
250 g
Catalog number 012544.09
also known as 012544-09
Price (USD)/ Each
74.65
Online exclusive
83.00 
Save 8.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
74.65
Online exclusive
83.00 
Save 8.35 (10%)
Add to cart
Indium(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number012544.09
Price (USD)/ Each
74.65
Online exclusive
83.00 
Save 8.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1312-43-2
IUPAC Namediindium(3+) trioxidandiide
Molecular FormulaIn2O3
InChI KeyPJXISJQVUVHSOJ-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[In+3].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Indium oxide is widely utilized as a n-type semiconductor, which is used in integrated circuits as a resistive element. It finds applications in batteries and as a part of some stain formulations. It is used as thin film coatings in optical, antistatic and infrared reflectors. It is doped with tin oxide and used as transparent conductive coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only