Thermo Scientific Chemicals

Indium(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012544.30
250 g, Each
Thermo Scientific Chemicals

Indium(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012544.30
250 g, Each
Quantity
Catalog number: 012544.30
also known as 012544-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1312-43-2
IUPAC Name
diindium(3+) trioxidandiide
Molecular Formula
In2O3
InChI Key
PJXISJQVUVHSOJ-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[In+3].[In+3]
Total Metal Impurities
0.01% max.

Description

Indium oxide is widely utilized as a n-type semiconductor, which is used in integrated circuits as a resistive element. It finds applications in batteries and as a part of some stain formulations. It is used as thin film coatings in optical, antistatic and infrared reflectors. It is doped with tin oxide and used as transparent conductive coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text