Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%, Thermo Scientific Chemicals
Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%, Thermo Scientific Chemicals

Silver plating
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012551.14
also known as 012551-14
Price (USD)/ Each
88.65
Online exclusive
98.20 
Save 9.55 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
98.20 
Save 9.55 (10%)
Add to cart
Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%, Thermo Scientific Chemicals
Catalog number012551.14
Price (USD)/ Each
88.65
Online exclusive
98.20 
Save 9.55 (10%)
-
Add to cart
Chemical Identifiers
CAS506-61-6
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Assay (unspecified)Ag 54.2% min
Silver platingPotassium silver cyanide is used in silver plating and in the manufacturing of antiseptics. It is a versatile bridging ligand and finds use for multidimensional polymer construction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only