Thermo Scientific Chemicals

Gold(I) potassium cyanide, Premion™, 99.96% (metals basis), Au 67.6% min, Thermo Scientific Chemicals

Catalog number: 012552.06
5 g, Each
Thermo Scientific Chemicals

Gold(I) potassium cyanide, Premion™, 99.96% (metals basis), Au 67.6% min, Thermo Scientific Chemicals

Catalog number: 012552.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 012552.06
also known as 012552-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13967-50-5
IUPAC Name
gold(1+) potassium bis(iminomethanide)
Molecular Formula
C2AuKN2
InChI Key
XTFKWYDMKGAZKK-UHFFFAOYSA-N
SMILES
[K+].[Au+].[C-]#N.[C-]#N
Total Metal Impurities
0.01% max.
Assay (unspecified)
Au 67.8% min

Description

Gold potassium cyanide is an important gold source used in electrolytic gold plating. The primary application of gold potassium cyanide is in the electroplating of metals. In addition, gold potassium cyanide is also used in the decorative plating of jewelry. It is useful for photoreduction of gold ions using the ZnO nanopowder. It is useful for the preparation of gold-gold junction electrode in the voltammetric glucose detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text