Thermo Scientific Chemicals

Molybdenum boride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012563.22
100 g, Each
Thermo Scientific Chemicals

Molybdenum boride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012563.22
100 g, Each
Quantity
Catalog number: 012563.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12006-98-3
IUPAC Name
molybdenioborane
Molecular Formula
BMo
InChI Key
LIGIGBHZRAABEF-UHFFFAOYSA-N
SMILES
[B+]=[Mo-]
Total Metal Impurities
1% max.

Description

Molybdenum borides have been used as hard coating materials for high temperature structural applications, and also in the form of MoB/CoCr cermet coatings. It is also used as a brazing material especially in electric components. Molybdenum boride has potential industrial applications such as abrasive, corrosion resistant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text