Thermo Scientific Chemicals

Hafnium silicide, Thermo Scientific Chemicals

Catalog number: 012564.09
10 g, Each
Thermo Scientific Chemicals

Hafnium silicide, Thermo Scientific Chemicals

Catalog number: 012564.09
10 g, Each
Quantity
Catalog number: 012564.09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12401-56-8
IUPAC Name
hafnium disilicon
Molecular Formula
HfSi2
InChI Key
TWRSDLOICOIGRH-UHFFFAOYSA-N
SMILES
[Si].[Si].[Hf]
Assay (unspecified)
99% min

Description

They have been used as ohmic contacts, Schottky barrier contacts, gate electrodes, local interconnects, and diffusion barriers. FeSi2 has become the remarkable source as high temperature thermoelectric material. It is also used as a powder in ceramic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text