Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Thermo Scientific Chemicals
Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Thermo Scientific Chemicals
Thermo Scientific™

Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012567.09
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS12069-85-1
IUPAC Namemethanidylidynehafniumylium
Molecular FormulaCHf
InChI KeyNVDNLVYQHRUYJA-UHFFFAOYSA-N
SMILES[C-]#[Hf+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Zr0.5% max.
Impurity contentZr <1%
It is used as an intermediate in chemical research, Pharmaceutical Intermediates,Organic Intermediates. Used in rocket nozzle, especially for re-entry space rocket nose cone.Due to its high hardness and melting point, HfC also widely use in hard alloy additives.Used in ceramic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only