Thermo Scientific Chemicals

Chromium boride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012570.14
25 g, Each
Thermo Scientific Chemicals

Chromium boride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012570.14
25 g, Each
Quantity
Catalog number: 012570.14
also known as 012570-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12007-16-8
IUPAC Name
diboron chromium
Molecular Formula
B2Cr
InChI Key
UHPOHYZTPBGPKO-UHFFFAOYSA-N
SMILES
[B].[B].[Cr]
Total Metal Impurities
1% max.

Description

Chromium boride,is been used in plasma technology and for high-performance electrodes in electron microscopes, metal matrix composites (MMCs), cutting tools and cermets for machining aluminum. chromium diboride is recognised as offering superior performance in corrosive-wear environments. Chromium diboride thin films possess desirable combinations of properties, which are attractive for a wide range of potential industrial applications. As a wear-resistant component of weld materials and powders for spraying in engineering and metallurgy, Protection of outer surfaces and bearing surfaces of machine parts against abrasive wear by plasma spraying, In aluminum evaporators manufacturing and for aluminum electrolyzes repair work, In nuclear engineering as neutron absorption material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text