Thermo Scientific Chemicals

Selenous acid, 97%, Thermo Scientific Chemicals

Catalog number: 012574.06
5 g, Each
Thermo Scientific Chemicals

Selenous acid, 97%, Thermo Scientific Chemicals

Catalog number: 012574.06
5 g, Each
Quantity
Catalog number: 012574.06
also known as 012574-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-00-8
IUPAC Name
selenous acid
Molecular Formula
H2O3Se
InChI Key
MCAHWIHFGHIESP-UHFFFAOYSA-N
SMILES
O[Se](O)=O
Assay (unspecified)
60.6% Se min

Description

As an oxidizing agent, as a reagent for alkaloidsSelenous acid is used in steel guns, wherein it changes the color of the steel from silver-grey to blue-grey. It serves as an important component of the Mecke reagent, which is utilized for drug testing. It acts as a source of selenium. It acts as a reagent for alkaloids and an oxidizing agent. Further, it is used in labeling radiopharmaceuticals. It is involved in the organic synthesis for the preparation of glyoxal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text