Palladium Silver foil, 0.127mm (0.005in) thick, 99.9% (metals basis excluding Pt), Thermo Scientific Chemicals
Palladium Silver foil, 0.127mm (0.005in) thick, 99.9% (metals basis excluding Pt), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Palladium Silver foil, 0.127mm (0.005in) thick, 99.9% (metals basis excluding Pt), Thermo Scientific Chemicals
Have Questions?
Quantity:
25 x 25 mm
Catalog number 012575.FF
also known as 012575-FF
Price (USD)/ Each
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialPalladium Silver foil
Name Note0.127mm (0.005 in.) thick
Recommended StorageAmbient temperatures
Avg. Mol. Wt. or Mol. Wt. RangePd:Ag; 75:25 wt%
TSCAYes