Platinum 30 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Platinum 30 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum 30 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
For ISA Type B Standard Grade Thermocouple
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 012576.BU
also known as 012576-BU
Price (USD)/ Each
-
Quantity:
25 cm
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPlatinum 30 wt% Rhodium wire
Name Note0.5mm (0.02 in.) dia.
Recommended StorageAmbient temperatures
Physical FormWire
Quantity25 cm
View more
Platinum 30 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple is used in the glass industry for furnace linings and complex fabrications. It is used as a catalyst in the preparation of nitric acid and in rayon spinnerets. It is used as a device for measuring high temperatures. It serves as a catalyst in the oxidation of the ammonia by atmospheric oxygen to produce nitric oxide, through Haber process.