Thermo Scientific Chemicals

Sodium selenite, anhydrous, 99% min, typically 99.75% min (metals basis), Thermo Scientific Chemicals

Catalog number: 012585.36
500 g, Each
Thermo Scientific Chemicals

Sodium selenite, anhydrous, 99% min, typically 99.75% min (metals basis), Thermo Scientific Chemicals

Catalog number: 012585.36
500 g, Each
Quantity
Catalog number: 012585.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-18-8
IUPAC Name
disodium selenite
Molecular Formula
Na2O3Se
InChI Key
BVTBRVFYZUCAKH-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-][Se]([O-])=O
Total Metal Impurities
1% max., typically 99.75% min

Description

In glass manufacturingSodium selenite is used in the manufacture of colorless glass. It acts as an ingredient in some food supplements. It is an animal feed used in pet foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text