Thermo Scientific Chemicals

Lithium metaborate, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012591.18
50 g, Each
Thermo Scientific Chemicals

Lithium metaborate, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012591.18
50 g, Each
Quantity
Catalog number: 012591.18
also known as 012591-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13453-69-5
IUPAC Name
tetraboron heptahydrate dilithium
Molecular Formula
B4H14Li2O7
InChI Key
DPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
Total Metal Impurities
0.1% max.

Description

Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text