Specifications
Chemical Name or MaterialLead
ColorGray
CAS7439-92-1
Health Hazard 1H350-H360FD-H362-H372
Health Hazard 2Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
View more