Potassium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals
Potassium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
1 kg
5 kg
Catalog number 012609.22
also known as 012609-22
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
Add to cart
Potassium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals
Catalog number012609.22
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
-
Add to cart
Chemical Identifiers
CAS584-08-7
IUPAC Namedipotassium carbonate
Molecular FormulaCK2O3
InChI KeyBWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa 0.005%
Impurity contentPO₄ 0.001%
Assay (unspecified)99.0% K₂CO₃ min
Impurity contentMg 0.002%
Heavy Metals(ICP-OES) 5ppm
View more
It may be used in pharmaceutical laboratories as a drying agent or as a source of potassium. It is also used in fire extinguishers, to make soap, glass and to soften water. Aqueous potassium carbonate plays an important role in carbon dioxide absorption promoted by piperazine. It is also essential for the absorption of carbon dioxide promoted by boric acid. It is also useful for the synthesis of azoles and diazines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only