Thermo Scientific Chemicals

Potassium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 012609.A7
5 kg, Each
Thermo Scientific Chemicals

Potassium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 012609.A7
5 kg, Each
Quantity
Catalog number: 012609.A7
also known as 012609-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
584-08-7
IUPAC Name
dipotassium carbonate
Molecular Formula
CK2O3
InChI Key
BWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]C([O-])=O
Impurity content
Ca 0.005%
Impurity content
PO₄ 0.001%
Assay (unspecified)
99.0% K₂CO₃ min
Impurity content
Mg 0.002%
Heavy Metals
(ICP-OES) 5ppm

Description

It may be used in pharmaceutical laboratories as a drying agent or as a source of potassium. It is also used in fire extinguishers, to make soap, glass and to soften water. Aqueous potassium carbonate plays an important role in carbon dioxide absorption promoted by piperazine. It is also essential for the absorption of carbon dioxide promoted by boric acid. It is also useful for the synthesis of azoles and diazines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text