Thermo Scientific Chemicals

Potassium chromate, ACS, 99.0% min

Catalog number: 012610.22
100 g, Each
Thermo Scientific Chemicals

Potassium chromate, ACS, 99.0% min

Catalog number: 012610.22
100 g, Each
Quantity
Catalog number: 012610.22
also known as 012610-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-00-6
IUPAC Name
dipotassium dioxochromiumbis(olate)
Molecular Formula
CrK2O4
InChI Key
XMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES
[K+].[K+].[O-][Cr]([O-])(=O)=O
Impurity content
Ca 0.005%
Impurity content
Cl 0.005%
Impurity content
Na 0.02%
Insoluble material
0.005%
pH
of a 5% solution 8.6-9.8 at 25?

Description

In finishing of leather, rust proofing of metals, textile mordant, as an analytical lab reagent. Potassium chromate enjoys a broad assortment of industrial purposes. In the fabric industry, manufacturers use potassium chromate to tan leather and dye clothing items and materials. In science, potassium chromate is often used as an indicator, meaning it can identify the presence, location and quantity of certain elements such as iron. Beyond these applications, potassium chromate also functions as an oxidizing agent in manufacturing and scientific experiments. Some nuclear power plants currently use substances other than potassium chromate to inhibit corrosion in closed cooling-water systems. Potassium chromate is applied as a chemical indicator of chlorine ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text