Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals
Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals
For Use As a Reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
1 kg
Catalog number 012613.06
also known as 012613-06
Price (USD)/ Each
30.65
Online exclusive
34.00 
Save 3.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
30.65
Online exclusive
34.00 
Save 3.35 (10%)
Add to cart
Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals
Catalog number012613.06
Price (USD)/ Each
30.65
Online exclusive
34.00 
Save 3.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13410-01-0
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.