Thermo Scientific Chemicals

Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt), Thermo Scientific Chemicals

Catalog number: 012627.NJ
∼0.5 g, Each
Thermo Scientific Chemicals

Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt), Thermo Scientific Chemicals

Catalog number: 012627.NJ
∼0.5 g, Each
Quantity
Catalog number: 012627.NJ
also known as 012627-NJ
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12033-81-7
IUPAC Name
dihydrogen dinitroplatinumbis(ylium) sulfate
Molecular Formula
H2N2O8PtS
InChI Key
SPPCQWREVYSZTJ-UHFFFAOYSA-N
SMILES
[H+].[H+].[O-]S([O-])(=O)=O.[O-][N+](=O)[Pt++][N+]([O-])=O
Concentration (weight/weight)
Pt 4-6%

Description

Dihydrogen dinitrosulfatoplatinate(II) solution is used in the preparation of electrodeposition of platinum films and biocompatible platinum thin film electrodeposition on tin substrates, which acts as the interconnects material for implantable medical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text