Thermo Scientific Chemicals

Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%, Thermo Scientific Chemicals

Catalog number: 012749.01
0.25 g, Each
Thermo Scientific Chemicals

Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%, Thermo Scientific Chemicals

Catalog number: 012749.01
0.25 g, Each
Quantity
Catalog number: 012749.01
also known as 012749-01
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12112-67-3
IUPAC Name
diiridium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
Molecular Formula
C16H24Cl2Ir2
InChI Key
ZFOUDQNHNLDNLD-MIXQCLKLSA-L
SMILES
[Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Assay (unspecified)
Ir 57.2% min

Description

Chloro(1,5-cyclooctadiene)iridium(I) dimer is widely used as a precursor to other iridium complexes, which finds application in homogeneous catalysis like carbonylation, hydrosilylation, hydrofomylation, asymmetric allylic substitutions, metathesis and chiral catalysis reactions. It is involved in the preparation of Crabtree's catalyst, which is used for hydrogenation and hydrogen-transfer reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text