Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%, Thermo Scientific Chemicals
Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.25 g
5 g
Catalog number 012749.03
also known as 012749-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS12112-67-3
IUPAC Namediiridium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
Molecular FormulaC16H24Cl2Ir2
InChI KeyZFOUDQNHNLDNLD-MIXQCLKLSA-L
SMILES[Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ir 57.2% min
Chloro(1,5-cyclooctadiene)iridium(I) dimer is widely used as a precursor to other iridium complexes, which finds application in homogeneous catalysis like carbonylation, hydrosilylation, hydrofomylation, asymmetric allylic substitutions, metathesis and chiral catalysis reactions. It is involved in the preparation of Crabtree's catalyst, which is used for hydrogenation and hydrogen-transfer reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only