Thermo Scientific Chemicals

Titanium(IV) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 012826.14
25 g, Each
Thermo Scientific Chemicals

Titanium(IV) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 012826.14
25 g, Each
Quantity
Catalog number: 012826.14
also known as 012826-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12039-13-3
IUPAC Name
disulfanylidenetitanium
Molecular Formula
S2Ti
InChI Key
CFJRPNFOLVDFMJ-UHFFFAOYSA-N
SMILES
S=[Ti]=S
Total Metal Impurities
0.2% max.

Description

Titanium(IV) sulfide is used in solid lubricant, cathode material in rechargeable batteries, corrosion resistance alloys, solid-state lithium batteries, hybrid electric vehicles and plug-in electric vehicles. It is used in the synthesis of nanoclusters, nanodisks and nanotubes. It has high electrical conductivity. Titanium electrodes are used in chloralkali electrolysis, electrodialysis, galvanic technology, light bulbs, and bone pins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text