Thermo Scientific Chemicals

Iron sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012842.09
10 g, Each
Thermo Scientific Chemicals

Iron sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012842.09
10 g, Each
Quantity
Catalog number: 012842.09
also known as 012842-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12068-85-8
IUPAC Name
disulfanylideneiron
Molecular Formula
FeS2
InChI Key
NFMAZVUSKIJEIH-UHFFFAOYSA-N
SMILES
S=[Fe]=S
Total Metal Impurities
0.1% max.

Description

Iron sulfide is used in photovoltaics and lithium ion battery cathodes. It is also studied as a potential material for the sequestration of environmental contaminants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text