Thermo Scientific Chemicals

Zinc oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 012855.14
25 g, Each
Thermo Scientific Chemicals

Zinc oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 012855.14
25 g, Each
Quantity
Catalog number: 012855.14
also known as 012855-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-13-2
IUPAC Name
oxozinc
Molecular Formula
OZn
InChI Key
XLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILES
O=[Zn]
Total Metal Impurities
0.0005% max.

Description

Zinc oxide is used as a sun screener. It is serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. Zinc oxide plays an important role in textiles as UV-absorber. It can protect rubber from fungi and UV light. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text