Thermo Scientific Chemicals

Titanium(II) hydride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012857.06
5 g, Each
Thermo Scientific Chemicals

Titanium(II) hydride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012857.06
5 g, Each
Quantity
Catalog number: 012857.06
also known as 012857-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7704-98-5
IUPAC Name
titanium(2+) dihydride
Molecular Formula
H2Ti
InChI Key
KAZWGWWZKAHTKC-UHFFFAOYSA-N
SMILES
[H-].[H-].[Ti++]
Appearance (Color)
Grey to black
Form
Powder
Assay from Suppliers CofA
Total metal impurities: ≤1.00% (ICP)
Particle Size
-325mesh

Description

Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text